Learn More
Hampton Research CRYSTAL SCREEN 2 NO44 185ML
SDP
Catalog No. 501503
Encompass
HR2-912-44/Crystal Screen 2 #44 (H08), 185 ml - 0.1 M Tris pH 8.5, 20% v/v Ethanol. Please note items from this supplier are non-returnable.
Catalog No. 50-150-3 Supplier Hampton Research Supplier No. HR291244
HR2-912-44/Crystal Screen 2 #44 (H08), 185 ml - 0.1 M Tris pH 8.5, 20% v/v Ethanol. Please note items from this supplier are non-returnable.