Learn More
Hampton Research CRYSTAL SCREEN 2 NO45 185ML
SDP
Catalog No. 501504
Encompass
HR2-912-45/Crystal Screen 2 #45 (H09), 185 ml - 0.01 M Nickel(II) chloride hexahydrate, 0.1 M Tris pH 8.5, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-150-4 Supplier Hampton Research Supplier No. HR291245
HR2-912-45/Crystal Screen 2 #45 (H09), 185 ml - 0.01 M Nickel(II) chloride hexahydrate, 0.1 M Tris pH 8.5, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.