Learn More
Hampton Research CRYSTAL SCREEN 2 NO46 185ML
SDP
Catalog No. 501505
Encompass
HR2-912-46/Crystal Screen 2 #46 (H10), 185 ml - 0.1 M Sodium chloride, 0.1 M BICINE pH 9.0, 20% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-150-5 Supplier Hampton Research Supplier No. HR291246
HR2-912-46/Crystal Screen 2 #46 (H10), 185 ml - 0.1 M Sodium chloride, 0.1 M BICINE pH 9.0, 20% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.