Learn More
Hampton Research CRYSTAL SCREEN 2 NO47 185ML
SDP
Catalog No. 501506
Encompass
HR2-912-47/Crystal Screen 2 #47 (H11), 185 ml - 0.1 M BICINE pH 9.0, 2.0 M Magnesium chloride hexahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-150-6 Supplier Hampton Research Supplier No. HR291247
HR2-912-47/Crystal Screen 2 #47 (H11), 185 ml - 0.1 M BICINE pH 9.0, 2.0 M Magnesium chloride hexahydrate. Please note items from this supplier are non-returnable.