Learn More
Hampton Research CRYSTAL SCREEN 2 NO48 185ML
SDP
Catalog No. 501507
Encompass
HR2-912-48/Crystal Screen 2 #48 (H12), 185 ml - 0.1 M BICINE pH 9.0, 2% v/v 1,4-Dioxane, 10% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.
Catalog No. 50-150-7 Supplier Hampton Research Supplier No. HR291248
HR2-912-48/Crystal Screen 2 #48 (H12), 185 ml - 0.1 M BICINE pH 9.0, 2% v/v 1,4-Dioxane, 10% w/v Polyethylene glycol 20,000. Please note items from this supplier are non-returnable.