Learn More
Hampton Research Crystal Screen Cryo, 10 mL, tube format
SDP
Catalog No. NC1755357
Encompass
Primary biased sparse matrix screen with cryo for proteins, soluble peptides, nucleic acids, & water soluble small molecules.
Catalog No. NC1755357 Supplier Hampton Research Supplier No. HR2122
Primary biased sparse matrix screen with cryo for proteins, soluble peptides, nucleic acids, & water soluble small molecules.