Learn More
Hampton Research Crystal Screen Cryo ; 10 ml, tube format
SDP
Catalog No. NC1672093
Encompass
Crystal Screen Cryo ; 10 ml, tube format, Cryo ready screening reagents for cryocrystallography, Sparse matrix formula efficiently samples salts, polymers, organics, & pH; HAMPTON RESEARCH mfg# HR2-133
Catalog No. NC1672093 Supplier Hampton Research Supplier No. HR2133
Add to Cart
Add to Cart
Crystal Screen Cryo ; 10 ml, tube format, Cryo ready screening reagents for cryocrystallography, Sparse matrix formula efficiently samples salts, polymers, organics, & pH; HAMPTON RESEARCH mfg# HR2-133