Learn More
Hampton Research CRYSTAL SCREEN CRYO NO01 185ML
SDP
Catalog No. 501508
Encompass
HR2-914-01/Crystal Screen Cryo #01 (A1), 185 ml - 0.02 M Calcium chloride dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-150-8 Supplier Hampton Research Supplier No. HR291401
HR2-914-01/Crystal Screen Cryo #01 (A1), 185 ml - 0.02 M Calcium chloride dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.