Learn More
Hampton Research CRYSTAL SCREEN CRYO NO02 185ML
SDP
Catalog No. 501509
Encompass
HR2-914-02/Crystal Screen Cryo #02 (A2), 185 ml - 0.26 M Potassium sodium tartrate tetrahydrate, 35% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-150-9 Supplier Hampton Research Supplier No. HR291402
HR2-914-02/Crystal Screen Cryo #02 (A2), 185 ml - 0.26 M Potassium sodium tartrate tetrahydrate, 35% v/v Glycerol. Please note items from this supplier are non-returnable.