Learn More
Hampton Research CRYSTAL SCREEN CRYO NO03 185ML
SDP
Catalog No. 501510
Encompass
HR2-914-03/Crystal Screen Cryo #03 (A3), 185 ml - 0.26 M Ammonium phosphate monobasic, 35% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-0 Supplier Hampton Research Supplier No. HR291403
HR2-914-03/Crystal Screen Cryo #03 (A3), 185 ml - 0.26 M Ammonium phosphate monobasic, 35% v/v Glycerol. Please note items from this supplier are non-returnable.