Learn More
Hampton Research CRYSTAL SCREEN CRYO NO04 185ML
SDP
Catalog No. 501511
Encompass
HR2-914-04/Crystal Screen Cryo #04 (A4), 185 ml - 0.075 M TRIS hydrochloride pH 8.5, 1.5 M Ammonium sulfate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-1 Supplier Hampton Research Supplier No. HR291404
HR2-914-04/Crystal Screen Cryo #04 (A4), 185 ml - 0.075 M TRIS hydrochloride pH 8.5, 1.5 M Ammonium sulfate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.