Learn More
Hampton Research CRYSTAL SCREEN CRYO NO05 185ML
SDP
Catalog No. 501512
Encompass
HR2-914-05/Crystal Screen Cryo #05 (A5), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-2 Supplier Hampton Research Supplier No. HR291405
HR2-914-05/Crystal Screen Cryo #05 (A5), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.