Learn More
Hampton Research CRYSTAL SCREEN CRYO NO06 185ML
SDP
Catalog No. 501513
Encompass
HR2-914-06/Crystal Screen Cryo #06 (A6), 185 ml - 0.16 M Magnesium chloride hexahydrate, 0.08 M TRIS hydrochloride pH 8.5, 24% w/v Polyethylene glycol 4,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-3 Supplier Hampton Research Supplier No. HR291406
HR2-914-06/Crystal Screen Cryo #06 (A6), 185 ml - 0.16 M Magnesium chloride hexahydrate, 0.08 M TRIS hydrochloride pH 8.5, 24% w/v Polyethylene glycol 4,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.