Learn More
Hampton Research CRYSTAL SCREEN CRYO NO07 185ML
SDP
Catalog No. 501514
Encompass
HR2-914-07/Crystal Screen Cryo #07 (A7), 185 ml - 0.07 M Sodium cacodylate trihydrate pH 6.5, 0.98 M Sodium acetate trihydrate, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-4 Supplier Hampton Research Supplier No. HR291407
HR2-914-07/Crystal Screen Cryo #07 (A7), 185 ml - 0.07 M Sodium cacodylate trihydrate pH 6.5, 0.98 M Sodium acetate trihydrate, 30% v/v Glycerol. Please note items from this supplier are non-returnable.