Learn More
Hampton Research CRYSTAL SCREEN CRYO NO08 185ML
SDP
Catalog No. 501515
Encompass
HR2-914-08/Crystal Screen Cryo #08 (A8), 185 ml - 0.14 M Sodium citrate tribasic dihydrate, 0.07 M Sodium cacodylate trihydrate pH 6.5, 21% v/v 2-Propanol, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-5 Supplier Hampton Research Supplier No. HR291408
HR2-914-08/Crystal Screen Cryo #08 (A8), 185 ml - 0.14 M Sodium citrate tribasic dihydrate, 0.07 M Sodium cacodylate trihydrate pH 6.5, 21% v/v 2-Propanol, 30% v/v Glycerol. Please note items from this supplier are non-returnable.