Learn More
Hampton Research CRYSTAL SCREEN CRYO NO09 185ML
SDP
Catalog No. 501516
Encompass
HR2-914-09/Crystal Screen Cryo #09 (A9), 185 ml - 0.17 M Ammonium acetate, 0.085 M Sodium citrate tribasic dihydrate pH 5.6, 25.5% w/v Polyethylene glycol 4,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-6 Supplier Hampton Research Supplier No. HR291409
HR2-914-09/Crystal Screen Cryo #09 (A9), 185 ml - 0.17 M Ammonium acetate, 0.085 M Sodium citrate tribasic dihydrate pH 5.6, 25.5% w/v Polyethylene glycol 4,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.