Learn More
Hampton Research CRYSTAL SCREEN CRYO NO11 185ML
SDP
Catalog No. 501518
Encompass
HR2-914-11/Crystal Screen Cryo #11 (A11), 185 ml - 0.07 M Sodium citrate tribasic dihydrate pH 5.6, 0.7 M Ammonium phosphate monobasic, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-8 Supplier Hampton Research Supplier No. HR291411
HR2-914-11/Crystal Screen Cryo #11 (A11), 185 ml - 0.07 M Sodium citrate tribasic dihydrate pH 5.6, 0.7 M Ammonium phosphate monobasic, 30% v/v Glycerol. Please note items from this supplier are non-returnable.