Learn More
Hampton Research CRYSTAL SCREEN CRYO NO12 185ML
SDP
Catalog No. 501519
Encompass
HR2-914-12/Crystal Screen Cryo #12 (A12), 185 ml - 0.18 M Magnesium chloride hexahydrate, 0.09 M HEPES sodium pH 7.5, 27% v/v 2-Propanol, 10% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-9 Supplier Hampton Research Supplier No. HR291412
HR2-914-12/Crystal Screen Cryo #12 (A12), 185 ml - 0.18 M Magnesium chloride hexahydrate, 0.09 M HEPES sodium pH 7.5, 27% v/v 2-Propanol, 10% v/v Glycerol. Please note items from this supplier are non-returnable.