Learn More
Hampton Research CRYSTAL SCREEN CRYO NO13 185ML
SDP
Catalog No. 501520
Encompass
HR2-914-13/Crystal Screen Cryo #13 (B1), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-152-0 Supplier Hampton Research Supplier No. HR291413
HR2-914-13/Crystal Screen Cryo #13 (B1), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M TRIS hydrochloride pH 8.5, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.