Learn More
Hampton Research CRYSTAL SCREEN CRYO NO14 185ML
SDP
Catalog No. 501521
Encompass
HR2-914-14/Crystal Screen Cryo #14 (B2), 185 ml - 0.19 M Calcium chloride dihydrate, 0.095 M HEPES sodium pH 7.5, 26.6% v/v Polyethylene glycol 400, 5% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-152-1 Supplier Hampton Research Supplier No. HR291414
HR2-914-14/Crystal Screen Cryo #14 (B2), 185 ml - 0.19 M Calcium chloride dihydrate, 0.095 M HEPES sodium pH 7.5, 26.6% v/v Polyethylene glycol 400, 5% v/v Glycerol. Please note items from this supplier are non-returnable.