Learn More
Hampton Research CRYSTAL SCREEN CRYO NO16 185ML
SDP
Catalog No. 501523
Encompass
HR2-914-16/Crystal Screen Cryo #16 (B4), 185 ml - 0.075 M HEPES sodium pH 7.5, 1.125 M Lithium sulfate monohydrate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-152-3 Supplier Hampton Research Supplier No. HR291416
HR2-914-16/Crystal Screen Cryo #16 (B4), 185 ml - 0.075 M HEPES sodium pH 7.5, 1.125 M Lithium sulfate monohydrate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.