Learn More
Hampton Research CRYSTAL SCREEN CRYO NO17 185ML
SDP
Catalog No. 501524
Encompass
HR2-914-17/Crystal Screen Cryo #17 (B5), 185 ml - 0.17 M Lithium sulfate monohydrate, 0.085 M TRIS hydrochloride pH 8.5, 25.5% w/v Polyethylene glycol 4,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-152-4 Supplier Hampton Research Supplier No. HR291417
HR2-914-17/Crystal Screen Cryo #17 (B5), 185 ml - 0.17 M Lithium sulfate monohydrate, 0.085 M TRIS hydrochloride pH 8.5, 25.5% w/v Polyethylene glycol 4,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.