Learn More
Hampton Research CRYSTAL SCREEN CRYO NO19 185ML
SDP
Catalog No. 501526
Encompass
HR2-914-19/Crystal Screen Cryo #19 (B7), 185 ml - 0.16 M Ammonium acetate, 0.08 M TRIS hydrochloride pH 8.5, 24% v/v 2-Propanol, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-152-6 Supplier Hampton Research Supplier No. HR291419
HR2-914-19/Crystal Screen Cryo #19 (B7), 185 ml - 0.16 M Ammonium acetate, 0.08 M TRIS hydrochloride pH 8.5, 24% v/v 2-Propanol, 20% v/v Glycerol. Please note items from this supplier are non-returnable.