Learn More
Hampton Research CRYSTAL SCREEN CRYO NO20 185ML
SDP
Catalog No. 501527
Encompass
HR2-914-20/Crystal Screen Cryo #20 (B8), 185 ml - 0.16 M Ammonium sulfate, 0.08 M Sodium acetate trihydrate pH 4.6, 20% w/v Polyethylene glycol 4,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-152-7 Supplier Hampton Research Supplier No. HR291420
HR2-914-20/Crystal Screen Cryo #20 (B8), 185 ml - 0.16 M Ammonium sulfate, 0.08 M Sodium acetate trihydrate pH 4.6, 20% w/v Polyethylene glycol 4,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.