Learn More
Hampton Research CRYSTAL SCREEN CRYO NO23 185ML
SDP
Catalog No. 501530
Encompass
HR2-914-23/Crystal Screen Cryo #23 (B11), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-153-0 Supplier Hampton Research Supplier No. HR291423
HR2-914-23/Crystal Screen Cryo #23 (B11), 185 ml - 0.2 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 30% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.