Learn More
Hampton Research CRYSTAL SCREEN CRYO NO24 185ML
SDP
Catalog No. 501531
Encompass
HR2-914-24/Crystal Screen Cryo #24 (B12), 185 ml - 0.14 M Calcium chloride dihydrate, 0.07 M Sodium acetate trihydrate pH 4.6, 14% v/v 2-Propanol, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-1 Supplier Hampton Research Supplier No. HR291424
HR2-914-24/Crystal Screen Cryo #24 (B12), 185 ml - 0.14 M Calcium chloride dihydrate, 0.07 M Sodium acetate trihydrate pH 4.6, 14% v/v 2-Propanol, 30% v/v Glycerol. Please note items from this supplier are non-returnable.