Learn More
Hampton Research CRYSTAL SCREEN CRYO NO25 185ML
SDP
Catalog No. 501532
Encompass
HR2-914-25/Crystal Screen Cryo #25 (C1), 185 ml - 0.07 M Imidazole pH 6.5, 0.7 M Sodium acetate trihydrate, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-2 Supplier Hampton Research Supplier No. HR291425
HR2-914-25/Crystal Screen Cryo #25 (C1), 185 ml - 0.07 M Imidazole pH 6.5, 0.7 M Sodium acetate trihydrate, 30% v/v Glycerol. Please note items from this supplier are non-returnable.