Learn More
Hampton Research CRYSTAL SCREEN CRYO NO26 185ML
SDP
Catalog No. 501533
Encompass
HR2-914-26/Crystal Screen Cryo #26 (C2), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-3 Supplier Hampton Research Supplier No. HR291426
Add to Cart
Add to Cart
HR2-914-26/Crystal Screen Cryo #26 (C2), 185 ml - 0.2 M Ammonium acetate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.