Learn More
Hampton Research CRYSTAL SCREEN CRYO NO27 185ML
SDP
Catalog No. 501534
Encompass
HR2-914-27/Crystal Screen Cryo #27 (C3), 185 ml - 0.14 M Sodium citrate tribasic dihydrate, 0.07 M HEPES sodium pH 7.5, 14% v/v 2-Propanol, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-4 Supplier Hampton Research Supplier No. HR291427
HR2-914-27/Crystal Screen Cryo #27 (C3), 185 ml - 0.14 M Sodium citrate tribasic dihydrate, 0.07 M HEPES sodium pH 7.5, 14% v/v 2-Propanol, 30% v/v Glycerol. Please note items from this supplier are non-returnable.