Learn More
Hampton Research CRYSTAL SCREEN CRYO NO28 185ML
SDP
Catalog No. 501535
Encompass
HR2-914-28/Crystal Screen Cryo #28 (C4), 185 ml - 0.17 M Sodium acetate trihydrate, 0.085 M Sodium cacodylate trihydrate pH 6.5, 25.5% w/v Polyethylene glycol 8,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-5 Supplier Hampton Research Supplier No. HR291428
HR2-914-28/Crystal Screen Cryo #28 (C4), 185 ml - 0.17 M Sodium acetate trihydrate, 0.085 M Sodium cacodylate trihydrate pH 6.5, 25.5% w/v Polyethylene glycol 8,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.