Learn More
Hampton Research CRYSTAL SCREEN CRYO NO29 185ML
SDP
Catalog No. 501536
Encompass
HR2-914-29/Crystal Screen Cryo #29 (C5), 185 ml - 0.065 M HEPES sodium pH 7.5, 0.52 M Potassium sodium tartrate tetrahydrate, 35% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-6 Supplier Hampton Research Supplier No. HR291429
HR2-914-29/Crystal Screen Cryo #29 (C5), 185 ml - 0.065 M HEPES sodium pH 7.5, 0.52 M Potassium sodium tartrate tetrahydrate, 35% v/v Glycerol. Please note items from this supplier are non-returnable.