Learn More
Hampton Research CRYSTAL SCREEN CRYO NO30 185ML
SDP
Catalog No. 501537
Encompass
HR2-914-30/Crystal Screen Cryo #30 (C6), 185 ml - 0.17 M Ammonium sulfate, 25.5% w/v Polyethylene glycol 8,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-7 Supplier Hampton Research Supplier No. HR291430
HR2-914-30/Crystal Screen Cryo #30 (C6), 185 ml - 0.17 M Ammonium sulfate, 25.5% w/v Polyethylene glycol 8,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.