Learn More
Hampton Research CRYSTAL SCREEN CRYO NO31 185ML
SDP
Catalog No. 501538
Encompass
HR2-914-31/Crystal Screen Cryo #31 (C7), 185 ml - 0.17 M Ammonium sulfate, 25.5% w/v Polyethylene glycol 4,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-8 Supplier Hampton Research Supplier No. HR291431
HR2-914-31/Crystal Screen Cryo #31 (C7), 185 ml - 0.17 M Ammonium sulfate, 25.5% w/v Polyethylene glycol 4,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.