Learn More
Hampton Research CRYSTAL SCREEN CRYO NO32 185ML
SDP
Catalog No. 501539
Encompass
HR2-914-32/Crystal Screen Cryo #32 (C8), 185 ml - 1.5 M Ammonium sulfate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-153-9 Supplier Hampton Research Supplier No. HR291432
HR2-914-32/Crystal Screen Cryo #32 (C8), 185 ml - 1.5 M Ammonium sulfate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.