Learn More
Hampton Research CRYSTAL SCREEN CRYO NO33 185ML
SDP
Catalog No. 501540
Encompass
HR2-914-33/Crystal Screen Cryo #33 (C9), 185 ml - 3.6 M Sodium formate, 10% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-0 Supplier Hampton Research Supplier No. HR291433
HR2-914-33/Crystal Screen Cryo #33 (C9), 185 ml - 3.6 M Sodium formate, 10% v/v Glycerol. Please note items from this supplier are non-returnable.