Learn More
Hampton Research CRYSTAL SCREEN CRYO NO34 185ML
SDP
Catalog No. 501541
Encompass
HR2-914-34/Crystal Screen Cryo #34 (C10), 185 ml - 0.07 M Sodium acetate trihydrate pH 4.6, 1.4 M Sodium formate, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-1 Supplier Hampton Research Supplier No. HR291434
HR2-914-34/Crystal Screen Cryo #34 (C10), 185 ml - 0.07 M Sodium acetate trihydrate pH 4.6, 1.4 M Sodium formate, 30% v/v Glycerol. Please note items from this supplier are non-returnable.