Learn More
Hampton Research CRYSTAL SCREEN CRYO NO35 185ML
SDP
Catalog No. 501542
Encompass
HR2-914-35/Crystal Screen Cryo #35 (C11), 185 ml - 0.075 M HEPES sodium pH 7.5, 0.6 M Sodium phosphate monobasic monohydrate, 0.6 M Potassium phosphate monobasic, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-2 Supplier Hampton Research Supplier No. HR291435
HR2-914-35/Crystal Screen Cryo #35 (C11), 185 ml - 0.075 M HEPES sodium pH 7.5, 0.6 M Sodium phosphate monobasic monohydrate, 0.6 M Potassium phosphate monobasic, 25% v/v Glycerol. Please note items from this supplier are non-returnable.