Learn More
Hampton Research CRYSTAL SCREEN CRYO NO36 185ML
SDP
Catalog No. 501543
Encompass
HR2-914-36/Crystal Screen Cryo #36 (C12), 185 ml - 0.065 M TRIS hydrochloride pH 8.5, 5.2% w/v Polyethylene glycol 8,000, 35% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-3 Supplier Hampton Research Supplier No. HR291436
HR2-914-36/Crystal Screen Cryo #36 (C12), 185 ml - 0.065 M TRIS hydrochloride pH 8.5, 5.2% w/v Polyethylene glycol 8,000, 35% v/v Glycerol. Please note items from this supplier are non-returnable.