Learn More
Hampton Research CRYSTAL SCREEN CRYO NO37 185ML
SDP
Catalog No. 501544
Encompass
HR2-914-37/Crystal Screen Cryo #37 (D1), 185 ml - 0.07 M Sodium acetate trihydrate pH 4.6, 5.6% w/v Polyethylene glycol 4,000, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-4 Supplier Hampton Research Supplier No. HR291437
HR2-914-37/Crystal Screen Cryo #37 (D1), 185 ml - 0.07 M Sodium acetate trihydrate pH 4.6, 5.6% w/v Polyethylene glycol 4,000, 30% v/v Glycerol. Please note items from this supplier are non-returnable.