Learn More
Hampton Research CRYSTAL SCREEN CRYO NO38 185ML
SDP
Catalog No. 501545
Encompass
HR2-914-38/Crystal Screen Cryo #38 (D2), 185 ml - 0.09 M HEPES sodium pH 7.5, 1.26 M Sodium citrate tribasic dihydrate, 10% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-5 Supplier Hampton Research Supplier No. HR291438
HR2-914-38/Crystal Screen Cryo #38 (D2), 185 ml - 0.09 M HEPES sodium pH 7.5, 1.26 M Sodium citrate tribasic dihydrate, 10% v/v Glycerol. Please note items from this supplier are non-returnable.