Learn More
Hampton Research CRYSTAL SCREEN CRYO NO39 185ML
SDP
Catalog No. 501546
Encompass
HR2-914-39/Crystal Screen Cryo #39 (D3), 185 ml - 0.085 M HEPES sodium pH 7.5, 1.7 M Ammonium sulfate, 1.7% w/v Polyethylene glycol 400, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-6 Supplier Hampton Research Supplier No. HR291439
HR2-914-39/Crystal Screen Cryo #39 (D3), 185 ml - 0.085 M HEPES sodium pH 7.5, 1.7 M Ammonium sulfate, 1.7% w/v Polyethylene glycol 400, 15% v/v Glycerol. Please note items from this supplier are non-returnable.