Learn More
Hampton Research CRYSTAL SCREEN CRYO NO40 185ML
SDP
Catalog No. 501547
Encompass
HR2-914-40/Crystal Screen Cryo #40 (D4), 185 ml - 0.095 M Sodium citrate tribasic dihydrate pH 5.6, 19% w/v Polyethylene glycol 4,000, 19% v/v 2-Propanol, 5% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-7 Supplier Hampton Research Supplier No. HR291440
HR2-914-40/Crystal Screen Cryo #40 (D4), 185 ml - 0.095 M Sodium citrate tribasic dihydrate pH 5.6, 19% w/v Polyethylene glycol 4,000, 19% v/v 2-Propanol, 5% v/v Glycerol. Please note items from this supplier are non-returnable.