Learn More
Hampton Research CRYSTAL SCREEN CRYO NO41 185ML
SDP
Catalog No. 501548
Encompass
HR2-914-41/Crystal Screen Cryo #41 (D5), 185 ml - 0.085 M HEPES sodium pH 7.5, 17% w/v Polyethylene glycol 4,000, 8.5% v/v 2-Propanol, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-8 Supplier Hampton Research Supplier No. HR291441
HR2-914-41/Crystal Screen Cryo #41 (D5), 185 ml - 0.085 M HEPES sodium pH 7.5, 17% w/v Polyethylene glycol 4,000, 8.5% v/v 2-Propanol, 15% v/v Glycerol. Please note items from this supplier are non-returnable.