Learn More
Hampton Research CRYSTAL SCREEN CRYO NO42 185ML
SDP
Catalog No. 501549
Encompass
HR2-914-42/Crystal Screen Cryo #42 (D6), 185 ml - 0.04 M Potassium phosphate monobasic, 16% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-154-9 Supplier Hampton Research Supplier No. HR291442
HR2-914-42/Crystal Screen Cryo #42 (D6), 185 ml - 0.04 M Potassium phosphate monobasic, 16% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.