Learn More
Hampton Research CRYSTAL SCREEN CRYO NO43 185ML
SDP
Catalog No. 501550
Encompass
HR2-914-43/Crystal Screen Cryo #43 (D7), 185 ml - 24% w/v Polyethylene glycol 1,500, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-0 Supplier Hampton Research Supplier No. HR291443
HR2-914-43/Crystal Screen Cryo #43 (D7), 185 ml - 24% w/v Polyethylene glycol 1,500, 20% v/v Glycerol. Please note items from this supplier are non-returnable.