Learn More
Hampton Research CRYSTAL SCREEN CRYO NO44 185ML
SDP
Catalog No. 501551
Encompass
HR2-914-44/Crystal Screen Cryo #44 (D8), 185 ml - 0.1 M Magnesium formate dihydrate, 50% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-1 Supplier Hampton Research Supplier No. HR291444
HR2-914-44/Crystal Screen Cryo #44 (D8), 185 ml - 0.1 M Magnesium formate dihydrate, 50% v/v Glycerol. Please note items from this supplier are non-returnable.