Learn More
Hampton Research CRYSTAL SCREEN CRYO NO45 185ML
SDP
Catalog No. 501552
Encompass
HR2-914-45/Crystal Screen Cryo #45 (D9), 185 ml - 0.16 M Zinc acetate dihydrate, 0.08 M Sodium cacodylate trihydrate pH 6.5, 14.4% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-2 Supplier Hampton Research Supplier No. HR291445
HR2-914-45/Crystal Screen Cryo #45 (D9), 185 ml - 0.16 M Zinc acetate dihydrate, 0.08 M Sodium cacodylate trihydrate pH 6.5, 14.4% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.