Learn More
Hampton Research CRYSTAL SCREEN CRYO NO46 185ML
SDP
Catalog No. 501553
Encompass
HR2-914-46/Crystal Screen Cryo #46 (D10), 185 ml - 0.16 M Calcium acetate hydrate, 0.08 M Sodium cacodylate trihydrate pH 6.5, 14.4% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-3 Supplier Hampton Research Supplier No. HR291446
HR2-914-46/Crystal Screen Cryo #46 (D10), 185 ml - 0.16 M Calcium acetate hydrate, 0.08 M Sodium cacodylate trihydrate pH 6.5, 14.4% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.