Learn More
Hampton Research CRYSTAL SCREEN CRYO NO49 185ML
SDP
Catalog No. 501556
Encompass
HR2-914-49/Crystal Screen Cryo #49, 185 ml - 0.8 M Lithium sulfate monohydrate, 1.6% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-6 Supplier Hampton Research Supplier No. HR291449
HR2-914-49/Crystal Screen Cryo #49, 185 ml - 0.8 M Lithium sulfate monohydrate, 1.6% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.