Learn More
Hampton Research CRYSTAL SCREEN CRYO NO50 185ML
SDP
Supplier:  Hampton Research HR291450
Encompass
HR2-914-50/Crystal Screen Cryo #50, 185 ml - 0.4 M Lithium sulfate monohydrate, 12% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-7